2-Methyl-4-cyanophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-2-methylphenylboronic acid, (4-Cyano-2-methylphenyl)boronic acid

A274837-250MG A274837-1G A274837-5G
103813-230EA 15 USD
103813-230 103853-158 76816-288 77092-994 77092-996
2-Methyl-4-cyanophenylboronic acid 95%
2-Methyl-4-cyanophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
MW: 160.966 g/mol
Boiling Pt: 376.1 °C (760 mm Hg)
Melting Pt: 287…305 °C
Flash Pt: 181.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750473
CAS Number: 313546-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR