4-Cyano-2-methoxybenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Cyano-2-methoxyphenyl)boronic acid

A118267-100MG A118267-250MG A118267-50MG A118267-1G
103799-020EA 35 USD
103799-020 103824-908 76822-264 76822-266 76822-268 77297-590
4-Cyano-2-methoxybenzeneboronic acid 97%
4-Cyano-2-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16618929
CAS Number: 1256345-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR