(4-Chloro-2-pyridinyl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: (4-Chloropyridin-2-yl)methanol, 4-Chloro-2-(hydroxymethyl)pyridine

A147302-100G A147302-10G A147302-1G A147302-25G
103803-758EA 9 USD
103803-758 103841-856 77162-600 77162-602 77317-222 77317-224
(4-Chloro-2-pyridinyl)methanol 98%
(4-Chloro-2-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.5709 g/mol
Boiling Pt: 238.6 °C (760 mm Hg)
Flash Pt: 98.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07437885
CAS Number: 63071-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR