4-Chlorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (4-Chlorophenyl)methanol

A116380-100G A116380-10G A116380-25G A116380-500G
103824-586EA 7 USD
103824-586 77108-264 77108-266 77108-268 77108-270
4-Chlorobenzyl alcohol 98%
4-Chlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 234 °C (760 mm Hg)
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004652
CAS Number: 873-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR