4-Chlorobenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (4-Chlorophenyl)methanamine

A101836-10G A101836-25G A101836-5G
103821-858EA 12 USD
103821-858 77148-240 77148-242 77282-844
4-Chlorobenzylamine 98%
4-Chlorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 215 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008121
CAS Number: 104-86-9
UN: 2735
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR