4-Chlorophenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chlorobenzeneboronic acid, Dihydroxy(4-chlorophenyl)borane, (4-Chlorophenyl)boronic acid, (4-Chlorophenyl)metaboric acid
A153651-100G
A153651-10G
A153651-1G
A153651-500G
A153651-1000G
A153651-5G
A153651-25G
103808-786EA
230
USD
103808-786
103842-720
77110-396
77110-398
77110-400
77110-402
77300-018
77300-020
77318-392
4-Chlorophenylboronic acid 98%
4-Chlorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₆BClO₂ Boiling Pt: 295.4 °C (760 mm Hg) Melting Pt: 284…289 °C Flash Pt: 132.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039137 CAS Number: 1679-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...