2-(4-chlorophenyl)acetaldehyde ≥95%

Supplier: Ambeed
Warning

Synonyms: (4-Chlorophenyl)acetaldehyde

A159105-250MG A159105-1G A159105-100MG
103805-224EA 151 USD
103805-224 103843-440 77163-136 77319-314 77892-086
2-(4-chlorophenyl)acetaldehyde ≥95%
2-(4-chlorophenyl)acetaldehyde

Precaución: Inert atmosphere, store in freezer, under –20 °C.

Formula: C₈H₇ClO
Boiling Pt: 235.8 °C (760 mm Hg)
Melting Pt: 87…88 °C
Flash Pt: 106.3 °C
Storage Temperature: Freezer
MDL Number: MFCD02261732
CAS Number: 4251-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR