(4-Chlorophenyl)(3,4-dihydroxyphenyl)methanone 95%

Supplier: Ambeed
Warning

A119660-10G A119660-1G A119660-5G
103799-814EA 11 USD
103799-814 76995-730 77311-924 77311-926
(4-Chlorophenyl)(3,4-dihydroxyphenyl)methanone 95%
(4-Chlorophenyl)(3,4-dihydroxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₃
MW: 248.66 g/mol
Boiling Pt: 468.8 °C (760 mm Hg)
Flash Pt: 237.3 °C
Storage Temperature: Ambient
MDL Number: MFCD08436199
CAS Number: 134612-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR