4-Chloro-3-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Chloro-3-methoxyphenyl)boronic acid

A219688-100MG A219688-10G A219688-250MG A219688-1G A219688-25G A219688-5G
103818-806EA 10 USD
103818-806 103849-684 76785-926 76785-928 76785-930 77088-236 77088-238 77088-240
4-Chloro-3-methoxyphenylboronic acid 98%
4-Chloro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.4000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318965
CAS Number: 89694-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR