4-Chloro-3-fluorobenzyl alcohol 97%

Supplier: Ambeed
Warning

A111035-10G A111035-25G A111035-1G A111035-5G
103797-780EA 9 USD
103797-780 103823-588 77094-000 77094-002 77284-078 77310-398
4-Chloro-3-fluorobenzyl alcohol 97%
4-Chloro-3-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 233.4 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00143292
CAS Number: 202925-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR