(4-Chloro-3-cyanophenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-cyanophenylboronic acid, 5-Borono-2-chlorobenzonitrile

A141150-10G A141150-250MG A141150-1G A141150-25G A141150-5G
103802-958EA 20 USD
103802-958 76782-936 76782-938 77316-094 77316-096 77316-098
(4-Chloro-3-cyanophenyl)boronic acid 98%
(4-Chloro-3-cyanophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClNO₂
MW: 181.382 g/mol
Boiling Pt: 380.4 °C (760 mm Hg)
Melting Pt: 300…304 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056360
CAS Number: 871332-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR