4-Chloro-2-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-methoxybenzeneboronic acid, (4-Chloro-2-methoxyphenyl)boronic acid

A288382-1G A288382-25G A288382-5G A288382-10G A288382-250MG
103813-792EA 8 USD
103813-792 103834-934 76837-494 76837-496 76837-498 77194-254 77194-256
4-Chloro-2-methoxyphenylboronic acid 98%
4-Chloro-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Boiling Pt: 345.4 °C (760 mm Hg)
Melting Pt: 128…134 °C
Flash Pt: 162.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02684317
CAS Number: 762287-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR