4'-Chlorobiphenyl-4-ylboronic acid 98%

Supplier: Ambeed
Warning

A184360-5G A184360-10G A184360-250MG
103815-748EA 9 USD
103815-748 103846-310 77083-922 77096-952 77096-954
4'-Chlorobiphenyl-4-ylboronic acid 98%
4'-Chlorobiphenyl-4-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BClO₂
Boiling Pt: 411 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08544389
CAS Number: 364044-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR