4-Butoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A220016-1G A220016-25G
103818-834EA 15 USD
103818-834 103849-716 77088-306 77088-308
4-Butoxyphenylboronic acid 98%
4-Butoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
Boiling Pt: 342.3 °C (760 mm Hg)
Melting Pt: 102…104 °C
Flash Pt: 160.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03427054
CAS Number: 105365-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR