(4-Bromothiazol-5-yl)methanol 95%

Supplier: Ambeed
Warning

A348817-100MG A348817-250MG A348817-1G A348817-5G
103826-000EA 57 USD
103826-000 103836-990 76788-356 76788-358 76840-072 76840-074
(4-Bromothiazol-5-yl)methanol 95%
(4-Bromothiazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₄BrNOS
MW: 194.0497 g/mol
Boiling Pt: 314.378 °C (760 mm Hg)
Flash Pt: 143.931 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909724
CAS Number: 262444-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR