(4-Bromo-2-pyridinyl)methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-2-(hydroxymethyl)pyridine, 4-Bromo-2-pyridinemethanol, (4-Bromopyridin-2-yl)methanol
A153615-10G
A153615-250MG
A153615-50MG
A153615-1G
A153615-5G
103804-536EA
13
USD
103804-536
103842-712
77150-482
77150-484
77150-486
77318-376
77318-378
(4-Bromo-2-pyridinyl)methanol 97%
(4-Bromo-2-pyridinyl)methanol
Precaución: for Research Use Only
Formula:
C₆H₆BrNO MW: 188.0200 g/mol Boiling Pt: 284.3 °C (760 mm Hg) Flash Pt: 125.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD09800557 CAS Number: 131747-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...