(4-Bromopyridin-2-yl)methanamine 95%
Supplier: Ambeed
|
Warning
|
A278632-250MG
A278632-100MG
103813-422EA
107
USD
103813-422
103853-364
77032-122
77668-226
(4-Bromopyridin-2-yl)methanamine 95%
(4-Bromopyridin-2-yl)methanamine
Precaución: for Research Use Only
Formula:
C₆H₇BrN₂ Boiling Pt: 262.8 °C (760 mm Hg) Storage Temperature: Freezer |
MDL Number:
MFCD07374903 CAS Number: 865156-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...