1-Bromo-4-(trimethylsilyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)trimethylsilane

A184781-100G A184781-10G A184781-1G A184781-25G A184781-5G
103815-782EA 6 USD
103815-782 103846-348 76825-050 76825-052 77083-968 77083-970 77083-972
1-Bromo-4-(trimethylsilyl)benzene 97%
1-Bromo-4-(trimethylsilyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₃BrSi
MW: 229.1890 g/mol
Boiling Pt: 230.7 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00094165
CAS Number: 6999-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR