4-Bromophenylhydrazinium chloride 95%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride

A115500-100G A115500-10G A115500-25G A115500-500G A115500-5G
103824-434EA 8 USD
103824-434 76810-128 76810-130 76810-132 76810-134 77297-376
4-Bromophenylhydrazinium chloride 95%
4-Bromophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₈BrClN₂
MW: 223.50 g/mol
Boiling Pt: 285.6 °C (760 mm Hg)
Melting Pt: 220…230 °C
Flash Pt: 126.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00012941
CAS Number: 622-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR