4-Bromophenylhydrazine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-hydrazinobenzene

A846456-10G A846456-5G
103841-222EA 30 USD
103841-222 76728-824 76845-470
4-Bromophenylhydrazine 98%
4-Bromophenylhydrazine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
Boiling Pt: 285.6 °C (760 mm Hg)
Melting Pt: 102…104 °C
Flash Pt: 126.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01935685
CAS Number: 589-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR