4-Bromo-N,N-diphenylaniline 98%

Supplier: Ambeed
Warning

A192471-100G A192471-10G A192471-500G A192471-25G A192471-5G
103847-050EA 5 USD
103847-050 76974-900 76974-902 76974-904 77302-290 77302-292
4-Bromo-N,N-diphenylaniline 98%
4-Bromo-N,N-diphenylaniline

Precaución: for Research Use Only

Formula: C₁₈H₁₄BrN
Boiling Pt: 419.1 °C (760 mm Hg)
Melting Pt: 108…112 °C
Flash Pt: 207.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01851266
CAS Number: 36809-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR