(4-Bromophenyl)acetaldehyde 95%

Supplier: Ambeed
Warning

A259992-5G A259992-250MG
103812-474EA 38 USD
103812-474 103852-356 77091-934 77612-486
(4-Bromophenyl)acetaldehyde 95%
(4-Bromophenyl)acetaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
MW: 199.0446 g/mol
Boiling Pt: 267.5 °C (760 mm Hg)
Melting Pt: 96…97 °C
Flash Pt: 99.676 °C
Storage Temperature: Freezer
MDL Number: MFCD02261728
CAS Number: 27200-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR