(4-Bromo-3-fluorophenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A146376-100MG A146376-10G A146376-250MG A146376-1G A146376-5G
103803-658EA 30 USD
103803-658 103841-746 77162-580 77162-582 77162-584 77299-514 77317-072
(4-Bromo-3-fluorophenyl)methanamine hydrochloride 97%
(4-Bromo-3-fluorophenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇BrFN·ClH
MW: 240.501 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546113
CAS Number: 1214342-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR