4-Bromo-2-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

A472333-100MG A472333-250MG A472333-1G A472333-25G A472333-5G
103828-044EA 24 USD
103828-044 76829-920 76829-922 76843-034 77886-223 77886-224
4-Bromo-2-methylphenylboronic acid 97%
4-Bromo-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16660270
CAS Number: 221006-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR