(4-Bromo-2-methoxyphenyl)acetonitrile 95%

Supplier: Ambeed
Warning

A100900-10G A100900-250MG A100900-25G A100900-1G A100900-5G
103795-682EA 13 USD
103795-682 103821-372 76780-410 76780-412 76780-414 77296-896 77296-898
(4-Bromo-2-methoxyphenyl)acetonitrile 95%
(4-Bromo-2-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO
Flash Pt: 140.32 °C
Storage Temperature: Ambient
MDL Number: MFCD11847504
CAS Number: 858523-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR