(4-Bromo-1,2-phenylene)dimethanol 97%

Supplier: Ambeed
Warning

A674868-1G A674868-25G A674868-5G
103840-518EA 129 USD
103840-518 76844-716 76844-718 77308-106
(4-Bromo-1,2-phenylene)dimethanol 97%
(4-Bromo-1,2-phenylene)dimethanol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.05 g/mol
Boiling Pt: 361.8±32.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD15147229
CAS Number: 171011-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR