(4-Boc-aminophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A298015-100G A298015-10G A298015-250MG A298015-1G A298015-25G A298015-5G A298015-500G
103814-170EA 9 USD
103814-170 103835-314 76816-696 76816-698 76816-700 76838-008 76838-010 76838-012 77672-512
(4-Boc-aminophenyl)boronic acid 98%
(4-Boc-aminophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₄
Melting Pt: 199…204 °C
Storage Temperature: Ambient
MDL Number: MFCD02093054
CAS Number: 380430-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR