4-Aminobenzyl alcohol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol
A125616-10G
A125616-1G
A125616-25G
A125616-5G
A125616-100G
103800-704EA
8
USD
103800-704
77136-684
77298-074
77298-076
77298-078
77549-492
4-Aminobenzyl alcohol 97%
4-Aminobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₉NO Melting Pt: 60…65 °C Storage Temperature: Freezer |
MDL Number:
MFCD00014782 CAS Number: 623-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...