(4-Aminophenyl)(morpholino)methanone 98%

Supplier: Ambeed
Warning

A110083-1G A110083-250MG
103797-614EA 43 USD
103797-614 103823-422 77283-936 77614-842
(4-Aminophenyl)(morpholino)methanone 98%
(4-Aminophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₂
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00158788
CAS Number: 51207-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR