(4-Aminobutyl)phosphonic acid 95%

Supplier: Ambeed
Warning

A159472-100MG A159472-250MG A159472-1G A159472-5G
103805-256EA 83 USD
103805-256 103843-476 77286-402 77286-404 77319-360 77319-362
(4-Aminobutyl)phosphonic acid 95%
(4-Aminobutyl)phosphonic acid

Precaución: for Research Use Only

Formula: C₄H₁₂NO₃P
Boiling Pt: 344.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00063426
CAS Number: 35622-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR