(4,6-Dichloro-3-pyridinyl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-5-(hydroxymethyl)pyridine, (4,6-Dichloropyridin-3-yl)methanol

A118084-250MG A118084-1G
103798-988EA 33 USD
103798-988 103824-880 77284-654 77297-580
(4,6-Dichloro-3-pyridinyl)methanol 95%
(4,6-Dichloro-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
Boiling Pt: 298.4 °C (760 mm Hg)
Flash Pt: 134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114262
CAS Number: 73998-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR