4-(Trimethylsilyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-(Trimethylsilyl)phenyl)boronic acid

A294038-1G A294038-25G A294038-5G A294038-100G A294038-10G A294038-250MG
103813-996EA 14 USD
103813-996 103835-136 76837-758 76837-760 76837-762 77250-728 77250-730 77250-732
4-(Trimethylsilyl)phenylboronic acid 95%
4-(Trimethylsilyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₅BO₂Si
Boiling Pt: 294.4 °C (760 mm Hg)
Melting Pt: 173…178 °C
Flash Pt: 131.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00093348
CAS Number: 17865-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR