4-(Trifluoromethyl)benzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Trifluoromethyl)phenyl)methanol, 4-(Hydroxymethyl)benzotrifluoride

A325373-25G A325373-5G A325373-100G A325373-10G A325373-500G
103836-342EA 5 USD
103836-342 76839-272 76839-274 76978-148 76978-150 76978-152
4-(Trifluoromethyl)benzyl alcohol 98%
4-(Trifluoromethyl)benzyl alcohol

Precaución: For research use only

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 213.7 °C (760 mm Hg)
Melting Pt: 18…20 °C
Flash Pt: 100.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004661
CAS Number: 349-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR