4-(Trifluoromethyl)phenylhydrazinium chloride 97%

Supplier: Ambeed
Warning

Synonyms: (4-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 4-(Trifluoromethyl)phenylhydrazine hydrochloride, 4-Hydrazinobenzotrifluoride hydrochloride, 1-(4-(Trifluoromethyl)phenyl)hydrazine hydrochloride

A109829-100G A109829-10G A109829-25G A109829-5G
103823-376EA 6 USD
103823-376 76765-736 76765-738 76765-740 77283-890
4-(Trifluoromethyl)phenylhydrazinium chloride 97%
4-(Trifluoromethyl)phenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₇H₈ClF₃N₂
Melting Pt: 200 °C
Flash Pt: 92.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00204233
CAS Number: 2923-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR