4-(Trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

A104094-100G A104094-10G A104094-1G A104094-25G A104094-500G A104094-5G
103822-364EA 14 USD
103822-364 77135-626 77135-628 77135-630 77135-632 77135-634 77309-222
4-(Trifluoromethyl)phenylboronic acid 98%
4-(Trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 245…250 °C
Flash Pt: 110.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR