4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (4-(Trifluoromethoxy)phenyl)hydrazine hydrochloride

A202839-100G A202839-10G A202839-25G A202839-5G A202839-1G
103817-418EA 8 USD
103817-418 103848-064 76770-214 76770-216 77086-176 77086-178 77302-830
4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClF₃N₂O
Melting Pt: 230 °C
Flash Pt: 195.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053033
CAS Number: 133115-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR