4-(trans-4-Propylcyclohexyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-(trans-4-propylcyclohexyl)phenyl)boronic acid

A130763-100G A130763-10G A130763-25G A130763-1G A130763-5G
103801-476EA 10 USD
103801-476 77271-984 77271-986 77271-988 77313-942 77313-944
4-(trans-4-Propylcyclohexyl)phenylboronic acid 95%
4-(trans-4-Propylcyclohexyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₂
MW: 246.1529 g/mol
Boiling Pt: 394.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06658197
CAS Number: 146862-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR