4-tert-Butylphenylhydrazine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (4-(tert-Butyl)phenyl)hydrazine hydrochloride

A132207-10G A132207-250MG A132207-1G A132207-25G A132207-5G
103801-702EA 9 USD
103801-702 76688-678 76688-680 77314-266 77314-268 77314-270
4-tert-Butylphenylhydrazine hydrochloride 95%
4-tert-Butylphenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇ClN₂
MW: 200.7084 g/mol
Melting Pt: 212…216 °C
Flash Pt: 137.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00013387
CAS Number: 128231-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR