4-tert-Butylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid

A503600-100G A503600-10G A503600-1G A503600-500G A503600-25G A503600-5G
103839-408EA 6 USD
103839-408 76774-510 76774-512 76774-514 76774-516 76843-384 76843-386
4-tert-Butylphenylboronic acid 98%
4-tert-Butylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₂
Melting Pt: 191…196 °C
Flash Pt: 133.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01009697
CAS Number: 123324-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR