4-(tert-Butoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butoxycarbonyl)benzeneboronic acid, (4-(tert-Butoxycarbonyl)phenyl)boronic acid, 4-tert-Butoxycarbonylphenylboronic acid

A170338-100MG A170338-250MG A170338-1G A170338-10G A170338-100G
103806-472EA 23 USD
103806-472 103844-746 76799-358 76799-360 77081-858 77567-496 77667-382
4-(tert-Butoxycarbonyl)phenylboronic acid 98%
4-(tert-Butoxycarbonyl)phenylboronic acid

Precaución: For research use only.

Formula: C₁₁H₁₅BO₄
Boiling Pt: 364.3 °C (760 mm Hg)
Melting Pt: 100…106 °C
Flash Pt: 174.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03411946
CAS Number: 850568-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR