(4-Pyrrolidin-1-ylphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-(Pyrrolidin-1-yl)phenyl)boronic acid

A312045-100MG A312045-250MG A312045-1G A312045-5G
103814-690EA 87 USD
103814-690 103835-880 77002-708 77002-710 77306-068 77306-070
(4-Pyrrolidin-1-ylphenyl)boronic acid 95%
(4-Pyrrolidin-1-ylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₂
MW: 191.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095137
CAS Number: 229009-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR