4-(Pyridin-4-yl)phenyl boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Pyridin-4-yl)phenyl)boronic acid

A179650-250MG A179650-1G A179650-25G A179650-10G
103807-452EA 23 USD
103807-452 103845-804 76985-714 77083-274 77301-622 77593-140
4-(Pyridin-4-yl)phenyl boronic acid 98%
4-(Pyridin-4-yl)phenyl boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD17170043
CAS Number: 1045332-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR