4-(Methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid

A397936-25G A397936-100G A397936-10G A397936-500G A397936-1G A397936-5G
103827-008EA 5 USD
103827-008 103838-048 76841-486 77252-088 77252-090 77252-092 77307-056 77307-058
4-(Methoxycarbonyl)phenylboronic acid 98%
4-(Methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Boiling Pt: 345.8 °C (760 mm Hg)
Melting Pt: 197…200 °C
Flash Pt: 162.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01632203
CAS Number: 99768-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR