4-Methoxycarbonyl-3,5-dimethylphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-(Methoxycarbonyl)-3,5-dimethylphenyl)boronic acid
A101028-100MG
A101028-250MG
A101028-1G
A101028-5G
103795-762EA
366
USD
103795-762
103821-454
77282-646
77282-648
77296-954
77296-956
4-Methoxycarbonyl-3,5-dimethylphenylboronic acid 97%
4-Methoxycarbonyl-3,5-dimethylphenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BO₄ Boiling Pt: 366.6±52.0 °C (760 mm Hg) Flash Pt: 175.503 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11111428 CAS Number: 876189-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...