4-(Bromomethyl)benzyl alcohol 97%

Supplier: Ambeed
Danger

Synonyms: [4-(Bromomethyl)phenyl]methanol, (4-(Bromomethyl)phenyl)methanol

A133543-100MG A133543-250MG A133543-1G A133543-5G
103801-882EA 31 USD
103801-882 77136-992 77136-994 77314-550 77314-552
4-(Bromomethyl)benzyl alcohol 97%
4-(Bromomethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
Melting Pt: 76…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275218
CAS Number: 71831-21-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR