4-(Bromomethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Bromomethyl)phenyl)metaboric acid, (4-(Bromomethyl)phenyl)boronic acid, Dihydroxy(4-(bromomethyl)phenyl)borane, 4-Bromomethylphenylboronic acid, 4-(Bromomethyl)benzeneboronic acid

A154746-10G A154746-1G A154746-25G A154746-5G
103804-676EA 5 USD
103804-676 103842-868 77190-892 77300-076 77300-078 77318-610
4-(Bromomethyl)phenylboronic acid 98%
4-(Bromomethyl)phenylboronic acid

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₇H₈BBrO₂
Boiling Pt: 343.1 °C (760 mm Hg)
Melting Pt: 78…81 °C
Flash Pt: 161.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318113
CAS Number: 68162-47-0
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR