(4-(Benzyloxy)phenyl)hydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A268201-1G A268201-25G A268201-5G A268201-100G A268201-10G
103812-904EA 5 USD
103812-904 103852-806 77092-554 77092-556 77092-558 77289-392 77289-394
(4-(Benzyloxy)phenyl)hydrazine hydrochloride 95%
(4-(Benzyloxy)phenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClN₂O
Melting Pt: 187…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137741
CAS Number: 52068-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR