4-(Benzyloxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxybenzeneboronic acid, (4-(Benzyloxy)phenyl)boronic acid

A113646-100G A113646-10G A113646-25G A113646-1G A113646-5G
103798-262EA 9 USD
103798-262 103824-104 76718-208 76718-210 76718-212 77297-256 77310-918
4-(Benzyloxy)phenylboronic acid 98%
4-(Benzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
Boiling Pt: 418.9 °C (760 mm Hg)
Melting Pt: 198…200 °C
Flash Pt: 207.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01075705
CAS Number: 146631-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR