4-Aminomethylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (4-Aminomethylphenyl)methanol, (4-(Aminomethyl)phenyl)methanol, 4-Aminomethylbenzyl alcohol, 4-(Aminomethyl)benzyl alcohol

A138471-100MG A138471-10G A138471-1G A138471-5G
103802-618EA 27 USD
103802-618 77162-222 77162-224 77315-648 77315-650
4-Aminomethylbenzyl alcohol 95%
4-Aminomethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
Melting Pt: 80…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06213766
CAS Number: 39895-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR