(4-(9-Phenyl-9H-carbazol-3-yl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A361777-1G A361777-5G A361777-100MG A361777-10G A361777-250MG
103826-288EA 23 USD
103826-288 103837-302 76840-516 76840-518 77195-356 77195-358 77195-360
(4-(9-Phenyl-9H-carbazol-3-yl)phenyl)boronic acid 97%
(4-(9-Phenyl-9H-carbazol-3-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₈BNO₂
MW: 363.22 g/mol
Boiling Pt: 555.9 °C (760 mm Hg)
Flash Pt: 290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD30063200
CAS Number: 1240963-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR